missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Vivaspin™ 20 Sample Concentrators
Shop All Cytiva Products
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
5 kDa
50 kDa
Unit Size:
Pack of 12
Descripción
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.
- Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
- Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
- Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
- Compatible pH range: 1 to 9
- Active membrane area: 6.0cm2
For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.
Especificaciones
Especificaciones
| Material (Membrane) | PES |
| MWCO | 30 kDa |
| Filtration Area | 6 cm2 |
| Quantity | 12/Pk. |
| Volume (Metric) | 5 to 20 mL |
| Length (Metric) | 116 mm |
| Width (Metric) | 30 mm |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido