missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Vivaspin™ 500 Sample Concentrators

Use for one-step, non-denaturing sample concentration in a single tube for minimal sample handling and reduced sample loss. Cytiva Vivaspin™ 500 Sample Concentrators cater to scientists working with sample volumes ranging from 100 μl to 20 ml, with a range of MWCO values from Mr 3 000 to 100 000.

Brand:  Cytiva 28-9322-36

Code : GE

Additional Details : Weight : 0.12400kg

 View more versions of this product

Product Code. 10554815

  • £107.13 / Pack of 25
Estimated Shipment: 16-07-2024
to see stock.
Add to basket

Description

Description

Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.

  • Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
  • Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
  • Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
  • Compatible pH range: 1 to 9
  • Active membrane area: 0.5cm2

For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.

Specifications

Specifications

PES
0.5 cm2
500 μL
11 mm
50 kDa
25/Pk.
50 mm
Product Suggestions

Product Suggestions

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote