Learn More
Sartorius Vivaspin™ 6
The Vivaspin™ 6 combines twin vertical membranes and thin flow channel design for high process speeds and concentration factors.
Brand: Sartorius VS0602IVD
Additional Details : Weight : 1.53100kg
Description
- CE certified according to EU 98/79/EC IVDD
- Easy-to-use centrifugal or solvent absorption devices
- No risk of sample cross-contamination
- Fast concentration with minimal membrane fouling
- High sample recoveries for increased sensitivity
Specifications
Ultrafiltration | |
PES | |
For in vitro diagnostics (IVD) applications. | |
Certified Quality |
Styrene butadiene copolymere | |
10 kDa | |
100 Pk. |