Learn More
Cytiva Vivaspin™ 6 Sample Concentrators
Use for one-step, non-denaturing sample concentration in a single tube for minimal sample handling and reduced sample loss. Cytiva Life Vivaspin™ 6 Sample Concentrators cater to scientists working with sample volumes ranging from 100 μl to 20 ml, with a range of MWCO values from Mr 3 000 to 100 000.
£222.91 - £471.00
Specifications
Material (Membrane) | PES |
---|---|
Filtration Area | 2.5 cm2 |
Quantity | 25/Pk. |
Volume (Metric) | 6 mL |
Length (Metric) | 122 mm |
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
10794217
|
Cytiva
28-9323-17 |
30 kDa |
£222.91
Pack of 25 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
10116914
|
Cytiva
28-9322-94 |
5 kDa |
£222.91
Pack of 25 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
10269574
|
Cytiva
28-9322-96 |
10 kDa |
£222.91
Pack of 25 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
10385021
|
Cytiva
VS0601 |
10 kDa |
£471.00
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10229624
|
Cytiva
28-9323-19 |
100 kDa |
£222.91
Pack of 25 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
10650561
|
Cytiva
28-9322-93 |
3 kDa |
£222.91
Pack of 25 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
10794407
|
Cytiva
28-9323-18 |
50 kDa |
£222.91
Pack of 25 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||||
Description
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.
- Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
- Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
- Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
- Compatible pH range: 1 to 9
- Active membrane area: 2.5cm2
For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.
Specifications
PES | |
25/Pk. | |
122 mm |
2.5 cm2 | |
6 mL | |
17 mm |