missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivaspin™ Turbo 15
The Vivaspin™ Turbo 15 is a 4 to 15 mL ultrafiltration unit with twin vertical PES and/or RC membranes and sleek internal design for fastest sample concentration with highest recoveries.
Brand: Sartorius VS15T01IVD
Additional Details : Weight : 0.45000kg
Description
- Vivaspin™ Turbo 15 PES and/or RC can handle up to 15 mL sample volume in swing bucket rotors and 11 mL in fixed angle rotors accepting 50 mL centrifuge tubes.
- The Vivaspin™Turbo 15's optimized design and sleek internal profile ensures maximum process speeds right down to the last few micro liters after 100 fold concentration.
- The UV joining technology allows for a smooth joint transition between membrane and plastic housing, allowing the collection of the complete concentrated sample into the unique pipette friendly dead stop pocket.
- Sleek internal design as well as large vertical twin membranes for ultra fast concentration
- High retentate recovery 95%
- pH 1 to 14 suited polystyrene housing with high chemical compatibility
- High flux PES membrane
- Universal rotor compatibility
- Safe sample concentration and easy recovery due to unique, angular and pipette friendly dead-stop pocket
- Volume graduation for easy detection / monitoring of the concentrate volume
- 100-fold concentration with twin vertical membranes
- Rotor Compatibility
- Please note: Vivaspin™ Turbo 15 (30 mm x 118 mm) is designed to fit into rotors that can accommodate Falcon 50 mL conical bottom tubes, e.g. Beckman Allegra 25R with TS-5.1-500 swing-out rotor with BUC 5 buckets and 368327 adaptors.
- Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors.
- Heraeus Multifuge 3 S-R with (Heraeus/Sorvall) 75006445 swing out rotor with 75006441 buckets and adaptors for Falcon 50 mL conical bottom tubes.
- These devices are not designed to fit into rotors that only accept round bottom 29 mm x 105 mm tubes, e.g. Sorvall SS34 or Beckmann JA 20.
- If your rotor accepts only 29 mm x 105 mm round bottom tubes, please use the Vivaspin™ 15, which can be used in either round bottom or conical centrifuge tubes.
Specifications
Ultrafiltration Filters and Devices | |
Polysulfone | |
Non-sterile | |
12 Pk. |
Styrene butadiene copolymere | |
10 kDa | |
For in vitro diagnostics (IVD) applications. | |
Certified Quality |