missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ Turbo 15 RC Ultrafiltration Centrifugal Concentrators
BIOPHARMA

Product Code. 16606952 Shop All Sartorius Products
Click to view available options
MWCO:
10 kDa
100 kDa
30 kDa
5 kDa
50 kDa
Quantity:
12/Pk.
48/Pk.
Unit Size:
Pack of 12
Pack of 48
This item is not returnable. View return policy

Product Code. 16606952

Brand: Sartorius VS15TR41

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Vivaspin™ Turbo 15 Regenerated Cellulose (RC) ultrafiltration centrifugal concentrators for protein, DNA, nanoparticle and virus concentration for 4-15 mL sample volumes. Acquire the fastest sample concentration with the highest recoveries for your protein samples.

Vivaspin™ Turbo centrifugal concentrators offer the optimal choice for maximum concentration or buffer exchange applications, with their broad range of MWCOs and market unique membrane choice of RC and PES membrane.

The Vivaspin™ Turbo 15 RC concentrators are the Regenerated Cellulose concentrators with the highest flow rates on the market due to their large, twin vertical membranes and sleek internal profile, which minimize protein polarization and subsequent fouling of the membrane and ensures maximum process speeds right down to the last 100 μL.

  • Unique optimal membrane choice (PES and RC)
  • Complete recoveries
  • Fastest concentrations
  • Highest chemical resistance
  • Easy retentate collection
  • Graduated and markable
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) Regenerated Cellulose
MWCO 100 kDa
Sample Volume 15 mL
Sterility Non-sterile
Housing Material Polypropylene
Quantity 12/Pk.
Type Centrifugal Concentrator
Volume (Metric) 15 mL
Length (Metric) 77 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.