Learn More
Sartorius Vivaspin™ Turbo 15 RC Ultrafiltration Centrifugal Concentrators
BIOPHARMA
Vivaspin™ Turbo 15 Regenerated Cellulose (RC) ultrafiltration centrifugal concentrators for protein, DNA, nanoparticle and virus concentration for 4-15 mL sample volumes. Acquire the fastest sample concentration with the highest recoveries for your protein samples.
Brand:  Sartorius VS15TR02
635.41 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 16636942
  • £783.00 / Pack of 48
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Vivaspin™ Turbo centrifugal concentrators offer the optimal choice for maximum concentration or buffer exchange applications, with their broad range of MWCOs and market unique membrane choice of RC and PES membrane.
The Vivaspin™ Turbo 15 RC concentrators are the Regenerated Cellulose concentrators with the highest flow rates on the market due to their large, twin vertical membranes and sleek internal profile, which minimize protein polarization and subsequent fouling of the membrane and ensures maximum process speeds right down to the last 100 μL.
  • Unique optimal membrane choice (PES and RC)
  • Complete recoveries
  • Fastest concentrations
  • Highest chemical resistance
  • Easy retentate collection
  • Graduated and markable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Regenerated Cellulose
Polypropylene
15 mL
10 kDa
48/Pk.
77 mm