Learn More
Sartorius Vivaspin™ Turbo 4
Vivaspin™ Turbo 4 offers unparalleled processing speeds right down to the last few microliters, thanks to larger, twin vertical membranes and sleek internal profiles. UV joining technology and Sartorius’ unique angular dead-stop picket allow easy, complete retrieval of the retentate sample.
Brand: Sartorius VS04T01IVD
Additional Details : Weight : 0.48400kg
Description
- CE certified according to EU 98/79/EC IVDD
- Easy-to-use centrifugal or solvent absorption devices
- No risk of sample cross-contamination
- Fast concentration with minimal membrane fouling
- High sample recoveries for increased sensitivity
- Typical Applications:
- Concentration of patient samples prior to electrophoresis or immuno-electrophoresis
- Concentration of urine, cerebrospinal fluid, or plasma to detect disease markers such as Bence Jones Protein
- Deproteinization of blood samples to aid detection of disease metabolites
Specifications
Ultrafiltration Filters and Devices | |
Polysulfone | |
Non-sterile | |
25 Pk. |
Styrene butadiene copolymere | |
10 kDa | |
For in vitro diagnostics (IVD) applications. | |
Certified Quality |