missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ Turbo 4 Ultrafiltration Unit
BIOPHARMA

Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.

Brand:  Sartorius VS04T31

261.36 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15311091

  • £322.00 / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.

  • High flux PES membrane
  • Retentate recovery >95% x pH 1-14 suited polystyrene housing with high chemical compatibility
  • Universal rotor compatibility
  • Volume graduation for easy detection / monitoring of the concentrate volume
  • UV joining technology allows for smooth joint transition between membrane and plastic housing, allowing collection of complete concentrated sample into unique pipette friendly dead stop pocket
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Turbo 4 Ultrafiltration Unit
50 kDa
Swing-out rotor : 4000 x gAngle rotor : 7500 x g
Sample type: Nanoparticles; Peptide; Protein
2 to 4 mL
PES
Concentration, Desalting, buffer exchange
25/Pk.
Centrifugal Unit
1225 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers