missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivaspin™ Turbo 4 Ultrafiltration Unit
Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.
Brand: Sartorius VS04T22
757.88 GBP valid until 2024-12-31
Use promo code "21930" to get your promotional price.
This item is not returnable.
View return policy
Description
Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.
- High flux PES membrane
- Retentate recovery >95% x pH 1-14 suited polystyrene housing with high chemical compatibility
- Universal rotor compatibility
- Volume graduation for easy detection / monitoring of the concentrate volume
- UV joining technology allows for smooth joint transition between membrane and plastic housing, allowing collection of complete concentrated sample into unique pipette friendly dead stop pocket
Specifications
Turbo 4 Ultrafiltration Unit | |
30 kDa | |
Swing-out rotor : 4000 x gAngle rotor : 7500 x g | |
Sample type: Nanoparticles; Peptide; Protein | |
2 to 4 mL |
PES | |
Concentration, Desalting, buffer exchange | |
100/Pk. | |
Centrifugal Unit | |
1225 mm |