missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ Turbo 4 Ultrafiltration Unit
BIOPHARMA

Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.

Brand:  Sartorius VS04T01

261.36 GBP valid until 2024-12-31
Use promo code "21930" to get your promotional price.


 View more versions of this product

Product Code. 16616752

  • £611.00 / Pack of 25
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.

  • High flux PES membrane
  • Retentate recovery >95% x pH 1-14 suited polystyrene housing with high chemical compatibility
  • Universal rotor compatibility
  • Volume graduation for easy detection / monitoring of the concentrate volume
  • UV joining technology allows for smooth joint transition between membrane and plastic housing, allowing collection of complete concentrated sample into unique pipette friendly dead stop pocket
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Turbo 4 Ultrafiltration Unit
10 kDa
Swing-out rotor : 4000 x gAngle rotor : 7500 x g
Sample type: Nanoparticles; Peptide; Protein
2 to 4 mL
PES
Concentration, Desalting, buffer exchange
25/Pk.
Centrifugal Unit
1225 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers