missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Vivid™ DBOMF Substrate

Product Code. 10347312
Click to view available options
Quantity:
0.1 mg
1 mg
Unit Size:
100µg
1mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10347312

Brand: Thermo Scientific™ P2979

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Vivid™ Substrates are blocked dyes that yield a minimal fluorescence signal until oxidation occurs.

Vivid™ Substrates are blocked dyes that yield a minimal fluorescence signal until oxidation occurs. Cytochrome P450 metabolism at either of two potential oxidation sites releases the highly fluorescent product. They have superior fluorescence, solubility, and kinetic properties compared to conventional fluorogenic probes. This results in higher sensitivity, greater signal-to-noise ratio, and better assay reproducibility. When used in combination with CYP450 BACULOSOMES™ Plus Reagents as part of the Vivid™ CYP450 Screening Kits, the Vivid™ Substrates enable rapid, robust detection of cytochrome P450 inhibitors. This particular substrate (Vivid ™ DBOMF Substrate) yields a product that emits a green fluorescence.

More About Vivid™ CYP450 Screening Kits

Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition. Vivid™ CYP450 Screening Kits include Vivid™ Substrate, Vivid™ Fluorescent Standard, reaction buffer, Vivid™ Regeneration System, NADP+, and CYP450 BACULOSOMES™ Plus Reagent. CYP450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells expressing a human CYP450 isozyme and human cytochrome P450 reductase. CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over human liver microsomes in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s.

Applications

  • High-throughput screening of enzyme-drug interactions
  • Compound profiling for drug inhibition of cytochrome P450 isozymes
  • Generation of predictive SAR models to guide compound acquisition
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Vivid™ CYP450 Screening kits include:
• Vivid™ CYP450 Fluorogenic substrates (also available separately)
• BACULOSOMES™ CYP450 reagents
• Fluorescent standards
• NADPH-regeneration system

Store reaction buffers at +4°C or room temperature. Store BACULOSOMES™, the NADPH-regeneration system, and NADP+ at -80°C. Store the Vivid™ substrates and standards at +4°C, protected from light. (For long-term storage, resuspend in organic solvent and keep at -20°C.)
Detection Method Fluorescence
For Use With (Application) Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target-based ADME/Tox assays
Product Line VIVID
Quantity 1 mg
Substrate DBOMF
Substrate Properties Chemical Substrate
Substrate Type Cytochrome P450 Substrate
Includes Vivid™ CYP450 fluorogenic substrates, BACULOSOMES™ CYP450 reagents, fluorescent standards, and an NADPH-regeneration system
Item Description Vivid™ DBOMF Substrate, 1mg, 572.6g/mol Molecular Weight, ≥ 97% Purity, Cytochrome P450 Substrate Type, Fluorescent Detection Method, Used For Cell Viability, Proliferation and Function, Pharma and Biopharma, Protein Assays and Analysis, Target-Based ADME/Tox Assays
Molecular Weight (g/mol) 572.6g/mol
Packaging 1 Each
Product Type Vivid™ DBOMF Substrate
Shipping Condition Approved for shipment on Wet or Dry Ice
Storage Requirements –20°C
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.