missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Vivid™ OOMR Substrate

Vivid™ Substrates are blocked dyes that yield a minimal fluorescence signal until oxidation occurs.

Brand:  Thermo Scientific™ P2868

 View more versions of this product

Product Code. 10768214

  • £134.00 / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Vivid™ Substrates are blocked dyes that yield a minimal fluorescence signal until oxidation occurs. Cytochrome P450 metabolism at either of two potential oxidation sites releases the highly fluorescent product. They have superior fluorescence, solubility, and kinetic properties compared to conventional fluorogenic probes. This results in higher sensitivity, greater signal-to-noise ratio, and better assay reproducibility. When used in combination with CYP450 BACULOSOMES™ Plus Reagents as part of the Vivid™ CYP450 Screening Kits, the Vivid™ Substrates enable rapid, robust detection of cytochrome P450 inhibitors. This particular substrate (Vivid ™ OOMR Substrate) yields a product that emits a red fluorescence.

More About Vivid™ CYP450 Screening Kits

Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition. Vivid™ CYP450 Screening Kits include Vivid™ Substrate, Vivid™ Fluorescent Standard, reaction buffer, Vivid™ Regeneration System, NADP+, and CYP450 BACULOSOMES™ Plus Reagent. CYP450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells expressing a human CYP450 isozyme and human cytochrome P450 reductase. CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over human liver microsomes in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s.

Applications

  • High-throughput screening of enzyme-drug interactions
  • Compound profiling for drug inhibition of cytochrome P450 isozymes
  • Generation of predictive SAR models to guide compound acquisition
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Vivid™ CYP450 Screening kits include:
• Vivid™ CYP450 Fluorogenic substrates (also available separately)
• BACULOSOMES™ CYP450 reagents
• Fluorescent standards
• NADPH-regeneration system

Store reaction buffers at +4°C or room temperature. Store BACULOSOMES™, the NADPH-regeneration system, and NADP+ at -80°C. Store the Vivid™ substrates and standards at +4°C, protected from light. (For long-term storage, resuspend in organic solvent and keep at -20°C.)
Liquid
VIVID™
OOMR
Cytochrome P450 Substrate
Vivid™ OOMR Substrate, 0.1mg, 355.4g/mol Molecular Weight, ≥ 97% Purity, Cytochrome P450 Substrate Type, Fluorescent Detection Method, Used For Cell Viability, Proliferation and Function, Pharma and Biopharma, Protein Assays and Analysis, Target-Based ADME/Tox Assays
1 Each
Approved for shipment on Wet or Dry Ice
Fluorescence
Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target-based ADME/Tox assays
0.1 mg
Chemical Substrate
Vivid™ CYP450 fluorogenic substrates, BACULOSOMES™ CYP450 reagents, fluorescent standards, and an NADPH-regeneration system
355.4g/mol
Vivid™ OOMR Substrate
–20°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.