Learn More
Poulten Graf™ Volac™ Borosilicate Glass Burettes with Glass Key
Featuring a flange at the top for ease of filling. Poulten Graf™ Volac™ Borosilicate Glass Burettes with Glass Key feature bold figures and fine graduation lines.
Brand:  Poulten Graf™ RB9803/NC/F
 View more versions of this product
Product Code. 10085871
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Removable glass jets are inscribed with instrument number for traceability
  • Flow time inscribed on the instrument
  • Certificate specifying accuracy at three points provided
  • Produced according to BS ISO 385
Specifications
Specifications
Blue
820 mm
0.05 mL
Straight
±0.03 mL
Class AS
25 mL
Borosilicate Glass
Interchangeable Glass Key in Glass Barrel
Glass
Volumetric Buret
BS 846, ISO 385