missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Volac™ Class B Soda Lime Glass Bulb Pipets

Brand:  Poulten Graf™ R370/K/2

Code : 01

Additional Details : Weight : 0.01000kg

Product Code. 12324368

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

BS1583, ISO648
Soda Lime Glass
2 Pack
Yellow
Bulb Pipet
B
Yes
±0.15 mL
100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers