missing translation for 'onlineSavingsMsg'
Learn More
Voriconazole, 98%, Thermo Scientific Chemicals Product Code.: 15442252

Voriconazole, 98%, Thermo Scientific Chemicals

Product Code. 15442252
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15442252

Brand: Thermo Scientific Acros 458670010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Voriconazole, CAS # 137234-62-9, is an antifungalagent that acts by inhibition of sterol biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Voriconazole is an antifungal compound that has activity against a wide variety of fungal species including Aspergillus and Candida
  • Voriconazole blocks sterol biosynthesis in Candida species through inhibition of CYP450-dependent 14-alpha sterol demethylase
  • It depletes the presence of ergosterol in the fungal cell membrane, which can lead to death or growth inhibition, depending on the organism

Applications

  • Voriconazole may be used in laboratory studies of drug safety, pharmacokinetics, and mechanism of action
TRUSTED_SUSTAINABILITY

Specifications

Type Voriconazole
CAS 137234-62-9
Melting Point 128.0°C to 134.0°C
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Quantity 1 g
Molecular Formula C16H14F3N5O
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in methanol,acetonitrile
InChI Key BCEHBSKCWLPMDN-MGPLVRAMSA-N
SMILES CC(C1=NC=NC=C1F)C(CN2C=NC=N2)(C3=C(C=C(C=C3)F)F)O
IUPAC Name (2R,3S)-2-(2,4-difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1,2,4-triazol-1-yl)butan-2-ol
Molecular Weight (g/mol) 349.317
PubChem CID 71616
ChEBI CHEBI:10023
Formula Weight 349.31
Percent Purity 98%
Chemical Name or Material Voriconazole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467291 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number458670010
Lot NumberA0466542 Certificate TypeCertificate of Analysis Date01/10/2024 Catalog Number458670010
Lot NumberA0463736 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number458670010
Lot NumberA0395401 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number458670010
Lot NumberA0410847 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number458670010
compliance-icons compliance-icons
Product Identifier
  • Voriconazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Voriconazole, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.