missing translation for 'onlineSavingsMsg'
Learn More

Voriconazole, Tocris Bioscience™

Product Code. 18752544
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Triazole antifungal agent

Triazole antifungal agent. Displays potent activity against B87964, Cryptococcus and Aspergillus species.

Chemical Identifiers

CAS 137234-62-9
Molecular Formula C16H14F3N5O
Molecular Weight (g/mol) 349.317
InChI Key BCEHBSKCWLPMDN-MGPLVRAMSA-N
Synonym voriconazole, vfend, 2r,3s-2-2,4-difluorophenyl-3-5-fluoropyrimidin-4-yl-1-1h-1,2,4-triazol-1-yl butan-2-ol, voriconazol, voriconazolum, voriconazole usan:inn:ban, unii-jfu09i87tr, vfend tn, 2r,3s-2-2,4-difluorophenyl-3-5-fluoropyrimidin-4-yl-1-1,2,4-triazol-1-yl butan-2-ol, vcz
PubChem CID 71616
ChEBI CHEBI:10023
IUPAC Name (2R,3S)-2-(2,4-difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1,2,4-triazol-1-yl)butan-2-ol
SMILES CC(C1=NC=NC=C1F)C(CN2C=NC=N2)(C3=C(C=C(C=C3)F)F)O

Specifications

Quantity 10 mg
Formula Weight 349.31
Percent Purity >99%
Chemical Name or Material Voriconazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'