Learn More
Fisherbrand™ Vortex Mixers Tube Holders
For use with Fisherbrand vortex mixers
Brand:  Fisherbrand™ 80863008
 View more versions of this product
Product Code. 15746959
  • £66.15 / Each
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Mount atop mixer for single-tube, hands-free mixing
  • Easily attach without tools
Specifications
Specifications
0.5 mL
9 x 9 in. (229 x 229 mm)
229 mm
229 mm
24
Microtube Holder
9 in.
9 in.
Vortex mixer