missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPARP Monoclonal Antibody (P193-4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA135746
This item is not returnable.
View return policy
Description
This gene encodes poly(ADP-ribosyl)transferase-like 1 protein, which is capable of catalyzing a poly(ADP-ribosyl)ation reaction. This protein has a catalytic domain which is homologous to that of poly (ADP-ribosyl) transferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly (ADP-ribosyl) transferase. Since this protein is not capable of binding DNA directly, its transferase activity may be activated by other factors such as protein-protein interaction mediated by the extensive carboxyl terminus.Specifications
VPARP | |
Monoclonal | |
0.25 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide | |
Q9UKK3 | |
PARP4 | |
Proprietary immunogen. | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Immunohistochemistry (Frozen), Western Blot | |
P193-4 | |
Unconjugated | |
PARP4 | |
193 kDa vault protein; ADP-ribosyltransferase (NAD+; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 1; ADP-ribosyltransferase diphtheria toxin-like 4; ADPRTL1; ARTD4; C030027K23Rik; E230037B21Rik; Gm743; H5 proline-rich; I-alpha-I-related; KIAA0177; p193; Parp4; PARP-4; PARPL; PARP-related; PARP-related/IalphaI-related H5/proline-rich; PH5P; poly (ADP-ribose) polymerase family, member 4; poly (ADP-ribose) polymerase)-like 1; poly [ADP-ribose] polymerase 4; poly(ADP-ribose) polymerase family member 4; poly(ADP-ribose) synthetase; poly(ADP-ribosyl)transferase-like 1; Protein mono-ADP-ribosyltransferase PARP4; Vault poly(ADP-ribose) polymerase; vault protein, 193-kDa; VAULT3; von Willebrand factor A domain containing 5C; VPARP; VWA5C | |
Mouse | |
purified | |
RUO | |
143 | |
4°, C | |
Liquid |