Learn More
Tocris Bioscience™ VTP 27999 trifluoroacetate
Highly potent and selective renin inhibitor
Brand: Tocris Bioscience™ 5563/10
This item is not returnable.
View return policy
Description
VTP 27999 trifluoroacetate is a highly potent and selective renin inhibitor (IC50 = 0.47 nM). Displays >1000-fold selectivity over related and unrelated off-targets. Blocks renin-stimulated ERK1/2 phosphorylation in vascular smooth muscle cells. Exhibits a different mode of action to aliskiren (Cat. No. 5491).Specifications
Methyl N-[2-[(R)-(3-Chlorophenyl)[(3R)-1-[[[(2S)-2-(methylamino)-3-[(3R)-tetrahydro-2H-pyran-3-yl]propyl]amino]carbonyl]-3-piperidinyl]methoxy]ethyl]carbamate trifluoroacetate | |
10 mg | |
C26H41ClN4O5.CF3CO2H | |
>97% |
1013937-63-7 | |
Additional Synthetic Protease Inhibitors | |
639.1 |