Learn More
Tocris Bioscience™ VU 0155069
Potent and selective PLD1 inhibitor
Brand:  Tocris Bioscience™ 3575/10
132.18 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18757497
  • £170.00 / 10mg
Explore more special offers
Description
Description
VU 0155069 is a potent and selective phospholipase D1 (PLD1) inhibitor (IC50 values are 46 and 933 nM for PLD1 and PLD2 respectively). Inhibits migration of human and mouse breast cancer cell lines in transwell assays.
Specifications
Specifications
>98%
10 mg
499.43
1781834-89-6
VU 0155069
VU 0155069