missing translation for 'onlineSavingsMsg'
Learn More

VUF 8430 dihydrobromide, Tocris Bioscience™

Product Code. 30001319
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, high affinity H4 agonist

High affinity (pKi = 7.5), potent histamine H4 receptor full agonist (pEC50 = 7.3). Displays moderate affinity for H3 receptors (pKi = 6.0) and weak partial agonist activity at H2 receptors.

Chemical Identifiers

CAS 100130-32-3
Molecular Formula C4H13Br2N5S
Molecular Weight (g/mol) 323.051
InChI Key GPWJSTKHQMIXCA-UHFFFAOYSA-N
Synonym vuf 8430 dihydrobromide, pseudourea, 2-2-guanidinoethyl-2-thio-, dihydrobromide, s-2-guanidylethyl-isothiourea, s,2-guanidinoethylisothiuronium bromide hydrobromide, carbamimidothioic acid, 2-aminoiminomethyl amino ethyl ester, dihydrobromide, 2-2-guanidinoethyl-2-thiopseudourea dihydrobromide, 2-aminoiminomethyl amino ethyl carbamimidothioic acid ester, carbamimidothioic acid,2-aminoiminomethyl amino ethyl ester, dihydrobromide 9ci, 2-carbamimidoylsulfanyl ethaneguanidine dihydrobromide, acmc-20m37r
PubChem CID 3063227
IUPAC Name 2-(diaminomethylideneamino)ethyl carbamimidothioate;dihydrobromide
SMILES C(CSC(=N)N)N=C(N)N.Br.Br

Specifications

Quantity 10 mg
Formula Weight 323.05
Percent Purity >98%
Chemical Name or Material VUF 8430 dihydrobromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'