missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Vybrant™ DyeCycle™ Violet Stain
Vybrant™ DyeCycle™ Violet Stain
Brand: Molecular Probes™ V35003
Code : 50
Additional Details : Weight : 0.00525kg
Description
Vybrant™ DyeCycle™ Violet Stain is a cell permeable DNA dye that can be used for cell cycle analysis and stem cell side population by flow cytometry.View a selection guide for all products related to cell cycle analysis of fixed and live cells in flow cytometry.
Precise
Successful cell cycle analysis requires a dye that is DNA selective and can stain cells in a homogeneous pattern minimizing fluorescence variability. The Vybrant™ DyeCycle™ Violet Stain is an ideal tool for DNA content analysis in living cells since the stain is cell permeable and, after binding double-stranded DNA, emits a fluorescent signal that is proportional to the DNA mass (see figure).
Low Cytotoxicity
Unlike stains that require high concentrations or have chemical structures that are toxic to cells, Vybrant™ DyeCycle™ stains exhibit relatively low cytotoxicity, allowing the possibility of sorting based on the phase of the cell cycle.
Minimal Compensation
Well-suited for the popular violet laser line, the Vybrant™ DyeCycle™ Violet Stain/DNA complex has fluorescence excitation and emission maxima of 369/437 nm, respectively (see figure).
The violet excitation and narrow emission of Vybrant™ DyeCycle™ Violet Stain make it ideal for multiplexing due to the limited spectral overlap with other common dyes (Alexa Fluor™ 488, FITC, and RPE) and fluorescent proteins (Green Fluorescent Protein (GFP) and mCherry) . Vybrant™ DyeCycle™ Violet Stain can also be used with UV excitation, having emission at ∼440 nm.
Flexible
Vybrant™ DyeCycle™ Violet Stain has been shown to not only work for both live cells and fixed cells in cell cycle assays, but to identify stem cell side populations and early progenitors in mammalian hematopoietic tissues (see figure).
Simple, Robust Staining Protocol
For cell analysis, simply prepare flow cytometry tubes each containing 1 mL of cells suspended in complete media at a cell concentration of 1 × 106 cells/mL. To each tube, add 1μL of Vybrant™ DyeCycle™ Violet Stain and mix well. Final stain concentration is 5μM. Incubate at 37°C for 30 minutes, protected from light. Keep cells at 37°C until acquisition. Analyze samples without washing or fixing on a flow cytometer using ∼405 nm excitation and ∼440 nm emission.
Specifications
5 mM | |
Fluorescence | |
Stain | |
369⁄437 | |
Tube(s) | |
200 Reactions | |
DMSO (Dimethylsulfoxide) |
Contains 1 vial of Vybrant™ DyeCycle™ violet stain (5 mM in DMSO). Store in refrigerator (2°C to 8°C) and protect from light. | |
Flow Cytometer | |
Vybrant™ DyeCycle™ Violet | |
Solution | |
DyeCycle™ | |
Room Temperature | |
Nucleic Acids |
For Research Use Only. Not for use in diagnostic procedures.