missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vybrant™ FLICA Caspase Apoptosis Assay Kits for flow cytometry

Product Code. p-7238533
Click to view available options
Label or Dye:
FAM, Hoechst 33342, Propidium Iodide
FAM, Hoechst 33344, Propidium Iodide
FMK, Hoechst 33342, Propidium Iodide
Description:
Vybrant FAM Caspase-3 and -7 Assay Kit
Vybrant FAM Caspase-8 Assay Kit
Vybrant FAM Poly Caspases Assay Kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10003342

Brand: Invitrogen™ V35117

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Detect apoptosis via flow cytometry with Vybrant FLICA Caspase Assays that detect poly and caspase-3/7/8 activities.

Quickly detect caspase-mediated apoptosis by flow cytometry with Vybrant FAM caspase assay kits for poly-caspases and caspases 3 and 7. The Vybrant FAM caspase assay kits use fluorescent inhibitor of caspases (FLICA) methodology to detect and report caspase activity. Specifically, these caspase assay kits contain engineered FLICA substrates FAM-VAD-FMK (for poly-caspases), FAM-DEVD-FMK (for caspase-3/7), and FAM-LETD-FMK (for caspase-8). They also include Hoechst 33342 and propidium iodide stains, enabling simultaneous evaluation of membrane permeability and cell cycle by flow cytometry.

The Vybrant FAM caspase assay kits for flow cytometry employ a novel approach to detecting active caspases: the kits take advantage of FLICA methodology, which is an affinity label. This affinity label associates a fluoromethyl ketone (FMK) moiety, which can react covalently with a cysteine, with a caspase-specific amino acid sequence. Different amino acid recognition sequences are used to detect different caspases: valine-alanine-aspartic acid (VAD) for poly caspases (including caspase-1, -3, -4, -5, -6, -7, -8, and -9), aspartic acid-glutamic acid-valine-aspartic acid (DEVD) for caspase-3/7, and leucine-glutamic acid-threonine-aspartic acid (LETD) for caspase-8. A carboxyfluorescein (FAM) group is attached as a reporter.

The FLICA reagent is thought to interact with the enzymatic reactive center of an activated caspase via the recognition sequence, and then to attach covalently through the FMK moiety. The FLICA inhibitor is cell permeant and noncytotoxic. Unbound FLICA molecules diffuse out of the cell and are washed away; the remaining fluorescent signal is a direct measure of the amount of active caspase present at the time the inhibitor was added. The approximate excitation and emission peaks of the FLICA, propidium iodide, and Hoechst 33342 reagents are 488 nm/⁄530 nm, 535⁄ nm/6617 nm, and 350 nm/⁄461 nm, respectively. The detection reagents included within the individual Vybrant FAM Poly Caspases Assay, Caspase-3 and -7 Assay, and Caspase-8 Assay apoptosis kits are 488⁄⁄530 FAM-VAD-FMK reagent, 488/⁄530 FAM-DEVD-FMK, and 488/⁄530 FAM-LETD-FMK, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Description Vybrant FAM Poly Caspases Assay Kit
Quantity 25 assays
Format Tube
Kit Contents 1 vial of FAM-VAD-FMK poly caspases reagent (FLICA reagent), 1 vial Hoechst 33342, 1 vial of propidium iodide, 1 vial of DMSO, 1 bottle of apoptosis fixative solution, and 1 bottle of apoptosis wash buffer (10X solution)
Product Type Caspase Assay Kit
Flow Cytometer Laser Lines UV, 488
Excitation/Emission FAM-VAD-FMDK: 488/530
PI: 535/617
Hoechst 33342: 350/461
Shipping Condition Room Temperature
Product Line Vybrant
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Label or Dye FMK, Hoechst 33342, Propidium Iodide
Label Type Other Label(s) or Dye(s)
Storage Requirements Store in refrigerator (2–8°C) and protect from light.
Show More Show Less
compliance-icons
Product Identifier
  • Vybrant FAM Poly Caspases Assay Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Dimethyl sulfoxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.