missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Vytac™ Formalin Disposal System, Vytac
Safely dispose of both 10% buffered and unbuffered formalin, including zinc formalin, using the Epredia™ Vytac™ Formalin Disposal System.
£181.00 - £185.00
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10240389
|
Epredia™
5401 |
Vytac; 1 Gal. (3.79L) |
£181.00
Pack of 4 |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10492659
|
Epredia™
5405 |
Vytac; 5 Gal. (18.9L) |
£185.00
Each |
Estimated Shipment: 19-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Disposal Solution:
- Breaks down formalin into inert components
- Neutralizes four parts formalin waste per one part Vytac
- When used as directed, neutralizes more than 99% of the waste formalin collected