missing translation for 'onlineSavingsMsg'
Learn More
Learn More
W-7 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
W-7 hydrochloride is a calmodulin antagonist. Inhibits Ca2+-calmodulin-dependent phosphodiesterase (IC50 = 28 μM) and myosin light chain kinase (IC50 = 51 μM).
Specifications
Specifications
CAS | 61714-27-0 |
Molecular Formula | C16H22Cl2N2O2S |
MDL Number | MFCD00012559 |
Quantity | 100 mg |
Synonym | w-7 hydrochloride, n-6-aminohexyl-5-chloro-1-naphthalenesulfonamide hydrochloride, w-7, hydrochloride, n-6-aminohexyl-5-chloronaphthalene-1-sulfonamide hydrochloride, c16h21cln2o2s hydrochloride, 1-naphthalenesulfonamide, n-6-aminohexyl-5-chloro-, monohydrochloride, n-6-aminohexyl-5-chloro-1-naphthalenesulfonamide monohydrochloride, n-6-aminohexyl-5-chloro-1-naphthalenesulfonamide hcl, n-6-aminohexyl-5-chloro-1-naphthalenesulfonamide, hcl, n-aminohexyl-5-chloro-1-napthalenesulfonamide hydrochloride |
InChI Key | OMMOSRLIFSCDBL-UHFFFAOYSA-N |
SMILES | [H+].[Cl-].NCCCCCCNS(=O)(=O)C1=C2C=CC=C(Cl)C2=CC=C1 |
IUPAC Name | hydrogen N-(6-aminohexyl)-5-chloronaphthalene-1-sulfonamide chloride |
Molecular Weight (g/mol) | 377.32 |
PubChem CID | 124887 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'