Description
- Facilitates the water purification and filtration process
- Aluminum and plastic construction with tap input
- Delivered with 2m of tube, screws and dowels
Specifications
65 x 11 cm | |
For Deionization Cartridges; Activated Carbon Cartridge; Micron Filter Cartridge; TOC Ion Exchange Cartridge; KCR Quick Coupling Kit | |
1 Each |
For water purification | |
Valve on the top of the support | |
Wall Bracket |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Fisherbrand™ Wall Bracket for Single Cartridge > For cartridge
Spot an opportunity for improvement?Share a Content Correction