missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Wall Mounted Arium pro Ultrapure Water System
GREENER_CHOICEBIOPHARMA

Provides widest selection of features and options for customised configuration.

Brand:  Sartorius™ H2OBASIC-B

Product Code. 11774996

  • £4930.00 / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Modular units pre-assembled according to specific applications
  • Lowest TOC level, along with real-time monitoring function
  • Efficient removal of endotoxins, DNase and RNase
  • High Flexibility
  • Total ease of operation
  • Meets and exceeds the ASTM Type 1 standard, ensuring reproducibility of results
  • Applications: HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC analysis, PCR, Electrophoresis, Endotoxin analysis, Immunocytochemistry, Pure water for preparation of cell culture media, Pure water for production of monoclonal antibodies, Photometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100μS/cm
Distilled, DI, RO, EDI
<5 ppb
No
0.2μm
<1 CFU/L
2 L/min.
Arium pro Ultrapure Water System
Wall
<1000ppb
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers