Learn More
Hughes Wall Mounted Indoor Unheated Safety Shower
Wall mounted unheated emergency safety shower, intended for use in environments where space is at a premium. This drench shower is suitable for use in all applications that don’t require an eyewash and are not subject to the possibility of freezing or overheating.
Brand: Hughes HSHH0023GS
This item is not returnable.
View return policy
Description
- Wall mounted unheated emergency safety shower, intended for use in environments where space is at a premium
- This drench shower is suitable for use in all applications that don’t require an eyewash and are not subject to the possibility of freezing or overheating
Specifications
Indoor Applications | |
Stainless Steel | |
Wall | |
Safety Shower |