Learn More
Brannan™ Spirit-filled Max Min Thermometer
Measures maximum and minimum temperaturess with non-toxic spirit filling. Brannan™ Spirit-filled Max Min Thermometer can be wall-mounted and resets with the push of a button.
Brand:  Brannan™ 63/096/0
Product Code. 13029212
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Measures maximum and minimum temperatures with non-toxic spirit filling.
  • Wall mounting in plastic case
  • 205mm long.
  • Push button reset.
Specifications
Specifications
205 mm
50°C
-30°C to +50°C
Non-toxic Spirit
-30°C