Learn More
3M™ Walnut Protein Rapid Kit L25WAL
Intended for screening for the presence of walnut proteins in clean-in-place (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
Brand:  3M™ 7100151246
Encompass
Product Code. 17273784
  • £440.40 / 25 tests
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Results visible at 11 ± 1 minute after application of the sample.
  • Rapid, qualitative immunochromatographic lateral flow test cartridges designed for the detection of allergenic food proteins.
  • Designed to detect both processed and unprocessed allergen proteins.
  • Test can accommodate environmental swab, CIP rinse water and food samples.
Specifications
Specifications
Microbiology Media
Allergen
25/Cs.