Learn More
R&L Enterprises™ Ward Pattern Furnace Tongs
Product Code. 11869701 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
590 mm
740 mm
Unit Size:
Each
Product Code. 11869701
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For protection and safety when handling heated objects in the laboratory. R&L Enterprises™ Ward Pattern Furnace Tongs are fitted with a circular shield to protect the hand against radiation from the furnace.
  • Manufactured from stainless steel
  • For handling a wide variety of crucibles, especially in muffle furnaces
  • Operated by thumb action with one hand
Specifications
For Use With (Application) For handling a wide variety of crucibles and to protect the hand against radiation from the furnace
Quantity 1 Ea.
Length (Metric) 740 mm
Material Stainless Steel
Type Furnace Tongs