Learn More
R&L Enterprises™ Ward Pattern Furnace Tongs
For protection and safety when handling heated objects in the laboratory. R&L Enterprises™ Ward Pattern Furnace Tongs are fitted with a circular shield to protect the hand against radiation from the furnace.
Brand:  R&L Enterprises™ 5704
 View more versions of this product
Product Code. 11899601
  • £271.00 / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from stainless steel
  • For handling a wide variety of crucibles, especially in muffle furnaces
  • Operated by thumb action with one hand
Specifications
Specifications
For handling a wide variety of crucibles and to protect the hand against radiation from the furnace
590 mm
Furnace Tongs
1 Ea.
Stainless Steel