missing translation for 'onlineSavingsMsg'
Learn More

SpotSee WarmMark™ Duo Ascending Time-Temperature Indicator

WarmMark™ Duo monitors temperatures during shipping and storage, indicating if goods experience unacceptable conditions. These reliable temperature indicators provide visual evidence of exposure to unacceptable temperature levels.

Brand:  SpotSee WD 10-34

Product Code. 17814086

  • £278.00 / Pack of 100
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Dual Threshold Time/Temperature indicator
  • Field-armable with pull tab
  • Cost-effective temperature monitor
Specifications

Specifications

REACH
Single-use
0.06 in.
98 mm
0.75 in.
Ascending time-temperature indicator
34°C
-30°C
-30°C to 34°C
Used in transport and storage of temperature sensitive products
3 days, 8 days, 14 days / 30 minutes
19 mm
98.1 x 19 x 1.5 mm
Pharmaceutical / Life Science Cold Chain Monitoring
1.5 mm
Acrylic Adhesive
3.88 in.
93°F
-22°F
-22 °F to 93 °F
Yes
± 1°C
Chemical Color Change
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SpotSee WarmMark™ Duo Ascending Time-Temperature Indicator > Time-temperature Indicator; 10°C and 34°C

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.