Learn More
Wash bottle round narrow neck hand trigger operated delivery 1.4mL per stroke HDPE 28mm R6 cap fitting natural 500mL
Product Code. 13121731
Click to view available options
Material:
HDPE
Unit Size:
Each
Product Code. 13121731
Brand: S Murray SAMCO P013/28+P513/14