missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Wash Buffer MBL3 for NucleoMag™ Blood
Additional wash buffer for use with NucleoMag™ Blood 200 μ/3 mL kits for magnetic-bead based isolation of genomic DNA from whole blood and biological fluids.
Brand: Macherey-Nagel™ 744848.1000
This item is not returnable.
View return policy
Description
NucleoMag™ Blood 200 μL/3 mL Kits
- Magnetic-bead based isolation of genomic DNA from whole blood
- One-tube procedure minimizes risk of cross-contamination
- Small elution volumes: > 50 μL (200 μL kit), > 1 mL (3 mL kit)
- Complete processing possible at room temperature
- Easily adapted to automated use or process manually
- Consistently high yields of genomic DNA
- Use whole blood, fresh or frozen, EDTA or citrate treated
- Downstream applications: PCR, real-time PCR, Southern blotting, any kind of enzymatic reaction
Specifications
Colorless | |
Ethanol; Sodium perchlorate; Water | |
1000 mL | |
Alcoholic | |
Liquid | |
Wash buffer MBL3 |
NucleoMag™ Blood 200 uL/3 mL kits | |
5.0 to 6.0 | |
64-17-5 | |
Room Temperature (18°C to 25°C) | |
Isolation of genomic DNA from whole blood |