missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wash Solution (concentrated), Thermo Scientific™
Wash Solution (concentrated)
Brand: Thermo Scientific R1621
45.00 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Additional Details : Weight : 0.19000kg
Description
Thermo Scientific GeneJET Wash Solution (concentrated) is a component of the GeneJET Plasmid Maxiprep Kit (K0491/K0492) and may be purchased separately. Provided in 180 mL aliquots.Specifications
Wash buffer | |
DNA and RNA purification and analysis, DNA extraction, plasmid DNA purification, midiprep | |
180 mL |
Wash Solution II, 180mL, Concentrated, Liquid, Wash Buffer, Used For DNA Extraction | |
Liquid | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.