missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Premiere Ecoforce™ Washroom Cleaner in Spray Bottle
Premiere Ecoforce™ Washroom Cleaner in Spray Bottle washes away the build up of dirt and grime that occurs with every day use, leaving behind a refreshing citrus fragrance.
Brand: Premiere Ecoforce™ 11516
Code : NEW
Additional Details : Weight : 0.00001kg
Description
- EU-Ecolabel accredited
- Citrus fragranced bathroom cleaner and descaler
- Rapidly removes watermarking and general soil
- Use for spray cleaning and damp wiping
- Suitable for use on all bathroom fittings, fixtures and surfaces
Specifications
Washroom Cleaner | |
100°C | |
Trigger Spray Bottle | |
6/Cs. |
3.5 | |
RTU | |
An effective all in one descaler and cleaner | |
750 mL |