Learn More
SCAT Waste Disposal Set, V2.0
The SCAT Waste Disposal Sets 1-4 are combinations of funnels and canisters for the safe disposal of liquid laboratory waste.
Brand:  SCAT 307315
285.48 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17132576
  • £382.00 / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of high quality and electrically conductive HDPE material
  • With grounding cable to dissipate the electrical charge
  • With splash guard and removable sieve for safe filling
  • Sets consist of ball or lid funnels for individual applications
  • 10 or 20 L disposal canisters with mechanical level indicators
  • Can be optionally combined with collection tray
All canisters and funnels are connected via an S60/61 thread and their components are perfectly matched to each other.
Specifications
Specifications
Funnel with lid MARCO, V2.0, S60/61, PE-HD electrostatic conductive, with removable sieve and earthing cable (318960), 10 L canister, S60/61, HDPE electrostatic conductive (107953), collecting tray with base insert (117985)
HPDE Electrostatic Conductive
S60/61
Set 2