Learn More
DWK Life Sciences Watch Glass Dish, made from soda-lime glass, fused rim
The concave shape of the watch glass dish allow working in very small quantities (e.g. for pretreatments) and covering laboratory glassware with wide openings.
£15.69 - £82.95
Specifications
Autoclavable | Not Autoclavable |
---|---|
Material | Soda Lime Glass |
For Use With (Application) | Weighing; Covering; Pretreatments; Drying of materials; Available in Boro 3.3 or soda-lime glass |
No. per Pack | 10/Pk. |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11786958
|
DWK Life Sciences
233214607 |
100 mm |
£57.02
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
10669891
|
DWK Life Sciences
233216608 |
250 mm |
£15.69
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
11776958
|
DWK Life Sciences
233214101 |
80 mm |
£49.90
Pack of 10 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
11796958
|
DWK Life Sciences
233215209 |
125 mm |
£81.01
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
15271839
|
DWK Life Sciences
233212409 |
40 mm |
£20.00
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
15281839
|
DWK Life Sciences
233213208 |
50 mm |
£29.21
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
15676925
|
DWK Life Sciences
233213405 |
60 mm |
£37.75
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
15686925
|
DWK Life Sciences
233213808 |
70 mm |
£25.28
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
15211849
|
DWK Life Sciences
233214307 |
90 mm |
£38.10
Pack of 10 |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
15221849
|
DWK Life Sciences
233215106 |
120 mm |
£60.45
Pack of 10 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
|||||
15231849
|
DWK Life Sciences
233215706 |
150 mm |
£55.80
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
15241849
|
DWK Life Sciences
233216102 |
200 mm |
£82.95
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Watch glasses, for example, are concave curved. The small trough created in this way allows liquids or solids to be filled in.
- Reduced risk of injury due to fused rim
Specifications
Not Autoclavable | |
Soda Lime Glass | |
Weighing; Covering; Pretreatments; Drying of materials; Available in Boro 3.3 or soda-lime glass | |
10/Pk. |