missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water with 0.1% ammonium acetate, Honeywell™
Click to view available options
Quantity:
2.5 L
Specifications
UN Number | NONH for all modes of transport |
Formula Weight | 18.02g/mol |
Quantity | 2.5 L |
Chemical Name or Material | Water with 0.1% ammonium acetate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'