missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water with 0.1% trifluoroacetic acid, Honeywell™
Click to view available options
Quantity:
2.5 L
Specifications
Specifications
CAS | 7732-18-5 |
Molecular Formula | H2O |
MDL Number | MFCD00011332 |
UN Number | NONH for all modes of transport |
Synonym | water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
SMILES | O |
IUPAC Name | oxidane |
Molecular Weight (g/mol) | 18.015 |
PubChem CID | 962 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'