missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, ACS Reagent, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
2.5 L
4 L
6 x 500 mL
4 x 2 L
4 x 4 L
Packaging:
Fiberboard Box
Glass Bottle
Poly Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
Melting Point | 0°C |
Boiling Point | 100°C |
UN Number | NONH for all modes of transport |
Beilstein | 2050024 |
Formula Weight | 18.02g/mol |
Grade | ACS Reagent |
Packaging | Fiberboard Box |
Density | 1.000g/mL at 3.98°C |
Quantity | 4 x 2 L |
Chemical Name or Material | Water |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction