missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
5 L
Packaging:
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Absorbance | Passes Test, 0.01 max. at 210nm, 0.005 max. at 254nm, 0.01 max. at 200nm, 0.005 max. at 300nm |
Color | White |
Boiling Point | 100°C |
Conductivity | 2μS/cm Max. |
Linear Formula | H2O |
Merck Index | 15,10237 |
Formula Weight | 18.02 |
Grade | ACS Reagent |
Packaging | Plastic Bottle |
Viscosity | 0.89 mPa.s (25°C) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'