missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water (Biotech Grade), Fisher BioReagents
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly CUBE
Description
Biotech-grade water can be used for several biological test procedures.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 7 |
Color | Colorless |
Melting Point | 0°C |
Boiling Point | 100°C |
Merck Index | 15, 10237 |
Formula Weight | 18.02 |
Grade | Biotech |
Packaging | Poly CUBE |
Vapor Pressure | 17.5mmHg at 20°C |
Filtered Through | Filtered through a 0.2-micron filter. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'