missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HyClone™ Water, Cell Culture Grade (Endotoxin-Free), Cytiva
6.21 GBP valid until 2024-07-31
Use promo code "21812" to get your promotional price.
Use promo code "21812" to get your promotional price.
Click to view available options
Quantity:
100 mL
500 mL
1 L
6 x 500 mL
6 x 1 L
Description
- Endotoxin-free; contains no detectable gram-negative bacteria
- Meets < 0.005 EU/mL endotoxin by turbidimetric assay
Specifications
pH | 5.0 to 7.0 |
Quantity | 100 mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'