Learn More
Water, Certified AR for Analysis, Certified AR, Fisher Chemical™
Brand: Fisher Chemical W/0100/21
46.68 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 22
Additional Details : CAS Number : 7732-18-5 Weight : 5.00000kg
Description
Water, Certified AR for Analysis, Certified AR
Specifications
HDPE plastic bottle | |
5 L |