missing translation for 'onlineSavingsMsg'
Learn More

Water, Certified AR for Analysis, Certified AR, Fisher Chemical™

Brand:  Fisher Chemical W/0100/FP21

Code : 21

Additional Details : CAS Number : 7732-18-5 Weight : 5.00000kg

 View more versions of this product

Product Code. 10501670

  • £51.55 / 5 L
In Stock
to see stock.
Add to basket

Description

Description

Water, Certified AR for Analysis, Certified AR

Specifications

Specifications

Colorless
100°C
11332
Plastic container
5 L
7732-18-5
H2O
18.02g/mol
17.5mmHg at 20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers