missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, CHROMASOLV™, for HPLC, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
4 L
6 x 1 L
Packaging:
Glass Bottle
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Absorbance | max. 0.005 at 400 nm, max. 0.005 at 250 nm, max. 0.01 at 210 nm, max. 0.01 at 205 nm |
Boiling Point | 100°C |
CAS Max % | 100.0000% |
Linear Formula | H2O |
Beilstein | 2050024 |
Formula Weight | 18.02 |
Grade | HPLC |
Packaging | Glass Bottle |
Density | 1.000 g/mL at 3.98 °C(lit.) |
Quantity | 2.5 L |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'