Learn More
Stuart™ Water Conversion Kit
Stuart™ Water Conversion Kit is specifically designed to improve the operation of Stuart™ Aquatron Water Stills
Brand: Stuart™ 99964-93
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- Laboratories typically prefer to feed their water stills with deionized or pre-treated water to obviate the need for boiling and descaling
- This kit easily converts Aquatron Water Stills into a deionized water feed
- The still is then permitted to operate with most types of deionizer, and also piped supplies of pre-treated water
Specifications
Aquatron Water Stills | |
Conversion to Deionised Water Feed | |
Water Conversion Kit |